Ethyl-4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate

A153837-1G A153837-250MG
77095-892EA 30.77 USD
77095-892 77095-894
Ethyl-4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate 95%
Ethyl 4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₃NO₃
MW: 285.2200 g/mol
MDL Number: MFCD00173369
CAS Number: 26893-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR