5,5'-Bis(trifluoromethyl)-2,2'-bipyridyl 98%

Supplier: Ambeed
Warning

Synonyms: 5,5'-Bis(trifluoromethyl)-2,2'-bipyridine

A153513-100MG A153513-1G A153513-250MG A153513-25G A153513-5G
77095-886EA 21.22 USD
77095-886 77095-888 77095-890 77685-268 77685-269
5,5'-Bis(trifluoromethyl)-2,2'-bipyridyl 98%
5,5'-Bis(trifluoromethyl)-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₂H₆F₆N₂
MW: 292.18 g/mol
MDL Number: MFCD24449717
CAS Number: 142946-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR