7-Methyl-1H-pyrazolo[4,3-c]pyridine 95%

Supplier: Ambeed
Warning

A153290-1G A153290-250MG
77095-880EA 1011.04 USD
77095-880 77095-882
7-Methyl-1H-pyrazolo[4,3-c]pyridine 95%
7-Methyl-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD13175928
CAS Number: 1049730-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR