Isonicotinic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid
A153232-1000G
A153232-100G
A153232-25G
A153232-500G
A153232-5G
77095-870EA
65.78
USD
77095-870
77095-872
77095-874
77095-876
77095-878
Isonicotinic acid 97%
Isonicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₅NO₂ MW: 123.11 g/mol |
MDL Number:
MFCD00006429 CAS Number: 55-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...