4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)phenol 98%
Supplier: Ambeed
|
Warning
|
A152273-1G
A152273-250MG
A152273-5G
77095-856EA
79.57
USD
77095-856
77095-858
77095-860
4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)phenol 98%
4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)phenol
Precaución: for Research Use Only
Formula:
C₁₁H₁₅BO₃ MW: 206.05 g/mol |
MDL Number:
MFCD09909077 CAS Number: 1192765-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...