1-(tert-Butoxycarbonyl)-4-ethylpiperidine-4-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A152096-100MG
A152096-1G
A152096-250MG
A152096-5G
77095-842EA
13.8
USD
77095-842
77095-844
77095-846
77095-848
1-(tert-Butoxycarbonyl)-4-ethylpiperidine-4-carboxylic acid 98%
1-(tert-Butoxycarbonyl)-4-ethylpiperidine-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₃H₂₃NO₄ |
MDL Number:
MFCD08062529 CAS Number: 188792-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...