2-Fluoro-6-nitroaniline 97%

Supplier: Ambeed
Warning

A151696-10G A151696-1G A151696-250MG A151696-25G A151696-5G
77095-832EA 20 USD
77095-832 77095-834 77095-836 77095-838 77095-840
2-Fluoro-6-nitroaniline 97%
2-Fluoro-6-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MW: 156.12
MDL Number: MFCD04112583
CAS Number: 17809-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR