4-Benzylmorpholin-3-one 95%

Supplier: Ambeed
Warning

A151412-1G A151412-250MG
77095-820EA 49.87 USD
77095-820 77095-822
4-Benzylmorpholin-3-one 95%
4-Benzylmorpholin-3-one

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.22 g/mol
MDL Number: MFCD10700061
CAS Number: 61636-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR