2'-Fluoro-4-pentyl-4''-propyl-1,1':4',1''-terphenyl 95%

Supplier: Ambeed
Warning

A151017-10G A151017-1G A151017-250MG
77095-810EA 99 USD
77095-810 77095-812 77095-814
2'-Fluoro-4-pentyl-4''-propyl-1,1':4',1''-terphenyl 95%
2'-Fluoro-4-pentyl-4''-propyl-1,1':4',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₆H₂₉F
MDL Number: MFCD11045061
CAS Number: 95759-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR