H-Pro-OEt hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: (S)-Ethyl pyrrolidine-2-carboxylate hydrochloride

A150169-1G A150169-25G
77095-788EA 7.43 USD
77095-788 77095-790
H-Pro-OEt hydrochloride 95%
H-Pro-OEt hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂·ClH
MW: 179.645 g/mol
MDL Number: MFCD00191004
CAS Number: 33305-75-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR