Azetidin-3-yl acetate 2,2,2-trifluoroacetate 98%

Supplier: Ambeed
Warning

A149436-100MG A149436-250MG A149436-25G A149436-5G
77095-780EA 24.41 USD
77095-780 77095-782 77095-784 77095-786
Azetidin-3-yl acetate 2,2,2-trifluoroacetate 98%
Azetidin-3-yl acetate 2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₇H₁₀F₃NO₄
MW: 229.15 g/mol
MDL Number: MFCD29917127
CAS Number: 1356114-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR