2-Ethynylnaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Ethynyl-naphthalene

A149319-100MG A149319-10G A149319-1G A149319-250MG A149319-25G A149319-5G A149319-100G
77095-768EA 5 USD
77095-768 77095-770 77095-772 77095-774 77095-776 77095-778 77901-846
2-Ethynylnaphthalene 98%
2-Ethynylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₈
MDL Number: MFCD00870424
CAS Number: 2949-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR