5-Amino-3-bromo-2-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-3-bromo-2-picoline

A148847-100G A148847-10G A148847-1G A148847-250MG A148847-25G A148847-5G
77095-756EA 462 USD
77095-756 77095-758 77095-760 77095-762 77095-764 77095-766
5-Amino-3-bromo-2-methylpyridine 98%
5-Amino-3-bromo-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.0372 g/mol
MDL Number: MFCD08060462
CAS Number: 186593-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR