1-Methyl-2-(methylthio)-5-nitro-1H-imidazole 96%
Supplier: Ambeed
|
Warning
|
A148699-100MG
A148699-250MG
77095-746EA
9.55
USD
77095-746
77095-748
1-Methyl-2-(methylthio)-5-nitro-1H-imidazole 96%
1-Methyl-2-(methylthio)-5-nitro-1H-imidazole
Precaución: for Research Use Only
Formula:
C₅H₇N₃O₂S MW: 173.19 g/mol |
MDL Number:
MFCD25959712 CAS Number: 1615-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...