3-Bromo-5-chloro-2-fluorobenzoic acid 95%

Supplier: Ambeed
Warning

A147937-1G A147937-250MG A147937-5G
77095-734EA 82 USD
77095-734 77095-736 77095-738
3-Bromo-5-chloro-2-fluorobenzoic acid 95%
3-Bromo-5-chloro-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MDL Number: MFCD21607478
CAS Number: 1269232-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR