2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine 95%

Supplier: Ambeed
Warning

Synonyms: 2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-D-cytidine, (2R,3R,4R,5R)-2-(4-Amino-5-fluoro-2-oxopyrimidin-1(2H)-yl)-5-methyltetrahydrofuran-3,4-diyl diacetate

A147683-100G A147683-1G A147683-250MG A147683-25G A147683-5G A147683-10G
77095-706EA 249.32 USD
77095-706 77095-708 77095-710 77095-712 77095-714 77685-239
2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine 95%
2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine

Precaución: for Research Use Only

Formula: C₁₃H₁₆FN₃O₆
MW: 329.28 g/mol
MDL Number: MFCD07369302
CAS Number: 161599-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR