2-Amino-3-iodo-5-nitropyridine 95%

Supplier: Ambeed
Warning

A147602-1G A147602-250MG A147602-5G A147602-10G
77095-700EA 11 USD
77095-700 77095-702 77095-704 77900-550
2-Amino-3-iodo-5-nitropyridine 95%
2-Amino-3-iodo-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄IN₃O₂
MDL Number: MFCD07368863
CAS Number: 25391-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR