D-(-)-Ornithine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Orn-OH·HCl, H-D-Orn-OH.HCl
A147590-100G
A147590-10G
A147590-1G
A147590-25G
A147590-5G
77095-690EA
264
USD
77095-690
77095-692
77095-694
77095-696
77095-698
D-(-)-Ornithine hydrochloride 98%
D-(-)-Ornithine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂N₂O₂·HCl |
MDL Number:
MFCD00012917 CAS Number: 16682-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...