Trelagliptin succinate 98+%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((6-(3-Aminopiperidin-1-yl)-3-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)-4-fluorobenzonitrile succinate

A147484-100MG A147484-10MG A147484-250MG A147484-50MG
77095-682EA 10 USD
77095-682 77095-684 77095-686 77095-688
Trelagliptin succinate 98+%
Trelagliptin succinate

Precaución: for Research Use Only

Formula: C₂₂H₂₆FN₅O₆
MW: 475.47 g/mol
MDL Number: MFCD22665720
CAS Number: 1029877-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR