5-Bromo-3-chloro-2-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

A147439-1G A147439-250MG
77095-678EA 26.53 USD
77095-678 77095-680
5-Bromo-3-chloro-2-fluorophenylboronic acid 97%
5-Bromo-3-chloro-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrClFO₂
MW: 253.26 g/mol
MDL Number: MFCD12026726
CAS Number: 1451393-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR