Cyclobutanecarboxamide 95%

Supplier: Ambeed
Warning

A147435-100MG A147435-1G A147435-250MG
77095-672EA 6.37 USD
77095-672 77095-674 77095-676
Cyclobutanecarboxamide 95%
Cyclobutanecarboxamide

Precaución: for Research Use Only

Formula: C₅H₉NO
MDL Number: MFCD00029076
CAS Number: 1503-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR