Ethyl 2,5-difluorobenzoate 97%

Supplier: Ambeed

A147249-100G A147249-1G A147249-250MG A147249-25G A147249-5G
77095-658EA 479 USD
77095-658 77095-660 77095-662 77095-664 77095-666
Ethyl 2,5-difluorobenzoate 97%
Ethyl 2,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MDL Number: MFCD06204339
CAS Number: 708-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR