4-Ethylsalicylaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 4-Ethyl-2-hydroxybenzaldehyde

A147221-1G A147221-250MG
77095-654EA 159.14 USD
77095-654 77095-656
4-Ethylsalicylaldehyde 95%
4-Ethylsalicylaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.175 g/mol
MDL Number: MFCD16997668
CAS Number: 161876-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR