[1,2,4]Triazolo[1,5-a]pyridin-6-ylboronic acid 95%

Supplier: Ambeed
Warning

A146968-1G A146968-250MG
77095-648EA 771.28 USD
77095-648 77095-650
[1,2,4]Triazolo[1,5-a]pyridin-6-ylboronic acid 95%
[1,2,4]Triazolo[1,5-a]pyridin-6-ylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BN₃O₂
MW: 162.94 g/mol
MDL Number: MFCD18250334
CAS Number: 1588769-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR