Z-Leu-OH·DCHA 97%

Supplier: Ambeed
Warning

Synonyms: Cbz-Leu-OH·DCHA, Z-Leu-OH.DCHA

A146444-100G A146444-25G
77095-642EA 46 USD
77095-642 77095-644 77095-646
Z-Leu-OH·DCHA 97%
Z-Leu-OH·DCHA

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄·C₁₂H₂₃N
MW: 446.6 g/mol
MDL Number: MFCD00065130
CAS Number: 53363-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR