4-Bromo-2-methoxy-5-methylaniline 97%

Supplier: Ambeed
Warning

A145850-5G A145850-1G A145850-250MG A145850-25G
77095-622EA 59 USD
77095-622 77901-876 77901-644 77901-005
4-Bromo-2-methoxy-5-methylaniline 97%
4-Bromo-2-methoxy-5-methylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MDL Number: MFCD12547798
CAS Number: 873980-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR