(2-Fluoro-4-iodo-3-pyridinyl)methanol 95%

Supplier: Ambeed
Warning

A145729-1G A145729-250MG A145729-5G
77095-616EA 16 USD
77095-616 77095-618 77095-620
(2-Fluoro-4-iodo-3-pyridinyl)methanol 95%
(2-Fluoro-4-iodo-3-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₅FINO
MDL Number: MFCD03095295
CAS Number: 171366-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR