Fraxinellone 95%

Supplier: Ambeed
Warning

Synonyms: (3R,3aR)-3-(Furan-3-yl)-3a,7-dimethyl-3a,4,5,6-tetrahydroisobenzofuran-1(3H)-one

A145557-1MG A145557-5MG
77095-612EA 21.22 USD
77095-612 77095-614
Fraxinellone 95%
Fraxinellone

Precaución: for Research Use Only

Formula: C₁₄H₁₆O₃
MW: 232.28 g/mol
MDL Number: MFCD11101451
CAS Number: 28808-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR