Sodium 2-((5-bromo-4-(4-cyclopropylnaphthalen-1-yl)-4H-1,2,4-triazol-3-yl)thio)acetate ≥98%

Supplier: Ambeed
Warning

Synonyms: Lesinurad sodium

A145508-10MG A145508-50MG A145508-5MG
77095-604EA 7 USD
77095-604 77095-606 77095-608 77095-610
Sodium 2-((5-bromo-4-(4-cyclopropylnaphthalen-1-yl)-4H-1,2,4-triazol-3-yl)thio)acetate ≥98%
Sodium 2-((5-bromo-4-(4-cyclopropylnaphthalen-1-yl)-4H-1,2,4-triazol-3-yl)thio)acetate

Precaución: for Research Use Only

Formula: C₁₇H₁₃BrN₃NaO₂S
MW: 426.26 g/mol
MDL Number: MFCD24444603
CAS Number: 1151516-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR