7-Chloro-2,3-dihydroimidazo[1,2-c]pyrimidin-5(1H)-one 95%

Supplier: Ambeed
Warning

A145103-100MG A145103-250MG
77095-578EA 53.05 USD
77095-578 77095-580
7-Chloro-2,3-dihydroimidazo[1,2-c]pyrimidin-5(1H)-one 95%
7-Chloro-2,3-dihydroimidazo[1,2-c]pyrimidin-5(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O
MW: 171.58 g/mol
MDL Number: MFCD25542223
CAS Number: 1421433-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR