4-Fluoronaphthalene-1,6-diol 80%

Supplier: Ambeed
Warning

A145054-100MG A145054-250MG
77095-574EA 169 USD
77095-574 77095-576
4-Fluoronaphthalene-1,6-diol 80%
4-Fluoronaphthalene-1,6-diol

Precaución: for Research Use Only

Formula: C₁₀H₇FO₂
MDL Number: MFCD24726538
CAS Number: 1824496-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR