3-Amino-2-methylpropanoic acid hydrate 95%

Supplier: Ambeed
Warning

A144949-10G A144949-1G A144949-5G
77095-568EA 200.52 USD
77095-568 77095-570 77095-572
3-Amino-2-methylpropanoic acid hydrate 95%
3-Amino-2-methylpropanoic acid hydrate

Precaución: for Research Use Only

Formula: C₄H₁₁NO₃
MW: 121.13 g/mol
MDL Number: MFCD00149561
CAS Number: 214139-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR