5-Methoxyisobenzofuran-1,3-dione 97%

Supplier: Ambeed
Warning

A144867-100MG A144867-10G A144867-1G A144867-250MG A144867-25G A144867-5G
77095-550EA 8.49 USD
77095-550 77095-552 77095-554 77095-556 77095-558 77095-560
5-Methoxyisobenzofuran-1,3-dione 97%
5-Methoxyisobenzofuran-1,3-dione

Precaución: for Research Use Only

Formula: C9H6O4
MW: 178.1415 g/mol
MDL Number: MFCD02670942
CAS Number: 28281-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR