3-Butylaniline 95%

Supplier: Ambeed
Warning

A144536-100MG A144536-1G A144536-250MG
77095-542EA 40 USD
77095-542 77095-544 77095-546
3-Butylaniline 95%
3-Butylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₅N
MW: 149.24
MDL Number: MFCD11845947
CAS Number: 5369-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR