3-Iodo-4-methoxyaniline hydrochloride 95%

Supplier: Ambeed
Warning

A144530-1G A144530-5G
77095-538EA 8.49 USD
77095-538 77095-540
3-Iodo-4-methoxyaniline hydrochloride 95%
3-Iodo-4-methoxyaniline hydrochloride

Precaución: for Research Use Only

Formula: C7H9ClINO
MW: 285.5099 g/mol
MDL Number: MFCD07780727
CAS Number: 261173-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR