BML-277 Chk2 inhibitor 98%

Supplier: Ambeed
Warning

A144417-10MG A144417-1MG A144417-50MG
77095-532EA 39.26 USD
77095-532 77095-534 77095-536
BML-277 Chk2 inhibitor 98%
BML-277 Chk2 inhibitor

Precaución: for Research Use Only

Formula: C₂₀H₁₄ClN₃O₂
MDL Number: MFCD08276917
CAS Number: 516480-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR