2,6-Dichloro-4-fluoroiodobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dichloro-5-fluoro-2-iodobenzene

A143251-100MG A143251-1G A143251-250MG
77095-508EA 41.38 USD
77095-508 77095-510 77095-512
2,6-Dichloro-4-fluoroiodobenzene 97%
2,6-Dichloro-4-fluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FI
MW: 290.889 g/mol
MDL Number: MFCD13194304
CAS Number: 939990-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR