3-Ethoxy-5-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

A142904-1G A142904-250MG
77095-496EA 23 USD
77095-496 77095-498
3-Ethoxy-5-(trifluoromethyl)aniline 97%
3-Ethoxy-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₉H₁₀F₃NO
MW: 205.17 g/mol
MDL Number: MFCD24038541
CAS Number: 1429903-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR