1-(4-Bromophenyl)naphthalene 97%

Supplier: Ambeed
Warning

A142591-1G A142591-250MG A142591-25G A142591-5G
77095-482EA 9.55 USD
77095-482 77095-484 77095-486 77095-488
1-(4-Bromophenyl)naphthalene 97%
1-(4-Bromophenyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₆H₁₁Br
MW: 283.17 g/mol
MDL Number: MFCD07369916
CAS Number: 204530-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR