5-(4-Hydroxy-3-methylphenyl)thiophene-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A142414-100MG A142414-1G A142414-250MG
77095-472EA 44.56 USD
77095-472 77095-474 77095-476
5-(4-Hydroxy-3-methylphenyl)thiophene-2-carbaldehyde 95%
5-(4-Hydroxy-3-methylphenyl)thiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂S
MW: 218.27 g/mol
MDL Number: MFCD18313179
CAS Number: 1261976-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR