H-D-Tyr(tBu)-OH 97%

Supplier: Ambeed
Warning

A142265-1G A142265-250MG
77095-468EA 22.28 USD
77095-468 77095-470
H-D-Tyr(tBu)-OH 97%
H-D-Tyr(tBu)-OH

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
MDL Number: MFCD00077112
CAS Number: 186698-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR