4-Methoxy-3-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-3-methylbenzeneboronic acid

A141652-1G A141652-5G A141652-25G
77095-454EA 7.43 USD
77095-454 77095-456 77566-850
4-Methoxy-3-methylphenylboronic acid 98%
4-Methoxy-3-methylphenylboronic acid

Precaución: For research use only.

Formula: C₈H₁₁BO₃
MW: 165.99 g/mol
MDL Number: MFCD02179464
CAS Number: 175883-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR