2'-Methylpropiophenone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Methylphenyl)propan-1-one, 2-(Ethylcarbonyl)toluene

A141141-1G A141141-250MG A141141-25G A141141-5G
77095-446EA 89.12 USD
77095-446 77095-448 77095-450 77095-452
2'-Methylpropiophenone 95%
2'-Methylpropiophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.202 g/mol
MDL Number: MFCD05663689
CAS Number: 2040-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR