trans-2-Carbomethoxycyclohexane-1-carboxyic acid 95%

Supplier: Ambeed
Warning

A140800-1G A140800-250MG A140800-25G A140800-5G
77095-430EA 205.82 USD
77095-430 77095-432 77095-434 77095-436
trans-2-Carbomethoxycyclohexane-1-carboxyic acid 95%
trans-2-Carbomethoxycyclohexane-1-carboxyic acid

Precaución: for Research Use Only

Formula: C₉H₁₄O₄
MW: 186.21 g/mol
MDL Number: MFCD01311243
CAS Number: 2484-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR