Ac-Nle-OH 97%

Supplier: Ambeed
Warning

Synonyms: N-Ac-Nle-OH, (S)-2-Acetamidohexanoic acid, (2S)-2-Acetamidohexanoic acid

A140667-10G A140667-25G A140667-5G
77095-424EA 59.42 USD
77095-424 77095-426 77095-428
Ac-Nle-OH 97%
Ac-Nle-OH

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MW: 173.2 g/mol
MDL Number: MFCD00151961
CAS Number: 15891-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR