Furo[3,2-c]pyridin-4(5H)-one 95%

Supplier: Ambeed
Warning

A139713-1G A139713-250MG A139713-5G
77095-410EA 26 USD
77095-410 77095-412 77592-652
Furo[3,2-c]pyridin-4(5H)-one 95%
Furo[3,2-c]pyridin-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₅NO₂
MDL Number: MFCD00183939
CAS Number: 26956-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR