5-Bromo-6-iodo-1H-indole 97%

Supplier: Ambeed
Warning

A139646-1G A139646-250MG
77095-406EA 366.02 USD
77095-406 77095-408
5-Bromo-6-iodo-1H-indole 97%
5-Bromo-6-iodo-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅BrIN
MW: 321.94 g/mol
MDL Number: MFCD09880052
CAS Number: 1000343-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR