5-Acetyl-2-amino-1,3-thiazole 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-ethanoyl-1,3-thiazole, 1-(2-Amino-1,3-thiazol-5-yl)ethan-1-one, 1-(2-Aminothiazol-5-yl)ethanone

A139515-100MG A139515-250MG A139515-1G A139515-5G
77095-398EA 13.8 USD
77095-398 77095-400 77592-644 77592-646
5-Acetyl-2-amino-1,3-thiazole 95%
5-Acetyl-2-amino-1,3-thiazole

Precaución: for Research Use Only

Formula: C₅H₆N₂OS
MW: 142.179 g/mol
MDL Number: MFCD00775044
CAS Number: 53159-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR