2-Amino-1-isopropylbenzimidazole 95%

Supplier: Ambeed
Warning

A139395-100MG A139395-250MG A139395-5G
77095-392EA 26.53 USD
77095-392 77095-394 77095-396
2-Amino-1-isopropylbenzimidazole 95%
2-Amino-1-isopropylbenzimidazole

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₃
MW: 175.23 g/mol
MDL Number: MFCD00159972
CAS Number: 90871-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR