1,4-Dimethylpiperazine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A139150-1G A139150-250MG
77095-372EA 55 USD
77095-372 77095-374
1,4-Dimethylpiperazine-2-carboxylic acid 95%
1,4-Dimethylpiperazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₄N₂O₂
MW: 158.19 g/mol
MDL Number: MFCD14583022
CAS Number: 58895-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR